Learn More
Fisherbrand™ Deuterium Lamps
Designed for compatibility with a variety of instruments. Fisherbrand™ Deuterium Lamps are individually tested, certified, and guaranteed to the same specifications as OEM.
Brand: Fisherbrand™ 11958-12
682.10 EUR valid until 2025-04-30
Use promo code "24163" to get your promotional price.
Description
- Produced in state of the art manufacturing facilities approved by lamp manufacturers
- Full traceability of manufactured products
- Lamps available for all major instruments
Specifications
UV | |
1 mm | |
1 Ea. |
400/160 nm | |
Hitachi L & U Series | |
Deuterium Lamp |
Promotions
-
Fisherbrand
Fisherbrand Sweet Deals on Top Products
Get up to 60% off select Fisherbrand productsPromo Code: 24163Expires: 2025-04-30