missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Deoxyribonuclease I, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
10mg
0.1g
Specifications
Specifications
Color | White |
MDL Number | MFCD00130918 |
SMILES | * |
IUPAC Name | Deoxyribonuclease |
Molecular Weight (g/mol) | 0.00 |
Quantity | 10mg |
Product Type | Deoxyribonuclease I |
Form | Powder |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Deoxyribonuclease I, MP Biomedicals™ >
Spot an opportunity for improvement?Share a Content Correction