Learn More
Fisherbrand™ Traceable™ Data Acquisition Software
Complete Data Acquisition Software enables measurement readings to be displayed and stored on a PC from up to 8 devices at a time.
Brand: Fisherbrand™ 3780413
This item is not returnable.
View return policy
Description
- Display and store measurement readings on a PC from up to 8 devices at a time
- Data capture runs in the background, enabling users to work on other programs simultaneously
- Set audible and visual notification alarms, or immediately send an email if an over/under condition occurs
- Software can be used to connect a meter directly to PC via cable
- Check software specification tables for meter compatibility
Specifications
Data Acquisition Software | |
Data Acquisition Software with USB Cable | |
37803-92, 37803-99, 91210-03, 19601-08 (Traceable™ 4132, 4310, 4137, 4169) |
No Display | |
Software CD and 6 ft. Cables (USB, Serial, and Instrument Connections) |
Product Content Correction
Product Title
Fisherbrand™ Traceable™ Data Acquisition Software > Data Acquisition Software with USB Cable