missing translation for 'onlineSavingsMsg'
Learn More
D-beta-Prolinol, 95%, Thermo Scientific Chemicals Product Code.: 15404268

D-beta-Prolinol, 95%, Thermo Scientific Chemicals

Product Code. 15404268
1 g
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15404268

Brand: Thermo Scientific Alfa Aesar H57871.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is an intermediate in organic syntheses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an intermediate in organic syntheses.

Solubility
Slightly soluble in water.

Notes
Hygroscopic. Store at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110013-18-8
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.149
MDL Number MFCD09607969
InChI Key QOTUIIJRVXKSJU-RXMQYKEDSA-N
Synonym r-pyrrolidin-3-ylmethanol, 3r-pyrrolidin-3-ylmethanol, 3r-pyrrolidin-3-yl methanol, r-3-hydroxymethyl pyrrolidine, d-beta-prolinol, 3-pyrrolidinemethanol, 3r, r-3-pyrrolidin-methanol, r-3-pyrrolidinemethanol, 3r-3-pyrrolidinemethanolShow MoreShow Less
PubChem CID 7446913
IUPAC Name [(3R)-pyrrolidin-3-yl]methanol
SMILES C1CNCC1CO

Specifications

Density 0.979
Boiling Point 173°C to 177°C
Quantity 1 g
Sensitivity Hygroscopic
Solubility Information Slightly soluble in water.
Formula Weight 101.15
Percent Purity 95%
Chemical Name or Material D-beta-Prolinol

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ19E021 Certificate TypeCertificate of Analysis Date15/01/2025 Catalog NumberH57871.03,H57871.MD
Lot NumberL06Y005 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH57871.03,H57871.MD
Lot NumberS15E039 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH57871.03,H57871.MD
Lot NumberZ19E021 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH57871.03,H57871.MD
Lot NumberS12A030 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH57871.03,H57871.MD

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
D-beta-Prolinol, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.