missing translation for 'onlineSavingsMsg'
Learn More
Cyclopropyl methyl ketone, 99%, Thermo Scientific Chemicals Product Code.: 11430014

Cyclopropyl methyl ketone, 99%, Thermo Scientific Chemicals

Product Code. 11430014
25 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11430014

Brand: Thermo Scientific Alfa Aesar A13540.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cyclopropyl methyl ketone was used in the preparation of homoallylic alcohols. Acetylcyclopropane is used as an intermediate for the manufacturing pharmaceuticals, agrochemicals and other organic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopropyl methyl ketone was used in the preparation of homoallylic alcohols. Acetylcyclopropane is used as an intermediate for the manufacturing pharmaceuticals, agrochemicals and other organic compounds.

Solubility
Fully miscible in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 765-43-5
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.118
MDL Number MFCD00001297
InChI Key HVCFCNAITDHQFX-UHFFFAOYSA-N
Synonym cyclopropyl methyl ketone, acetylcyclopropane, methyl cyclopropyl ketone, ethanone, 1-cyclopropyl, 1-cyclopropyl-ethanone, 1-cyclopropylethan-1-one, ketone, cyclopropyl methyl, cyclopropylmethylketone, unii-n27yy1xcfh, cyclopropylethanoneShow MoreShow Less
PubChem CID 13004
IUPAC Name 1-cyclopropylethanone
SMILES CC(=O)C1CC1

Specifications

Melting Point <-70°C
Density 0.898
Boiling Point 113°C to 114°C
Flash Point 16°C (61°F)
Refractive Index 1.424
Quantity 25 g
UN Number UN1992
Beilstein 1633817
Solubility Information Fully miscible in water.
Formula Weight 84.12
Percent Purity 99%
Chemical Name or Material Cyclopropyl methyl ketone
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10245586 Certificate TypeCertificate of Analysis Date01/02/2023 Catalog NumberA13540.14,A13540.36,A13540.22
Lot Number10233915 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13540.14,A13540.36,A13540.22
Lot Number10192071 Certificate TypeCertificate of Analysis Date07/08/2020 Catalog NumberA13540.14,A13540.36,A13540.22
Lot Number10225563 Certificate TypeCertificate of Analysis Date07/08/2020 Catalog NumberA13540.14,A13540.36,A13540.22
Lot Number10223400 Certificate TypeCertificate of Analysis Date07/08/2020 Catalog NumberA13540.14,A13540.36,A13540.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cyclopropyl methyl ketone, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.