missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CUTC Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA558469
This item is not returnable.
View return policy
Description
Immunogen sequence: KRARIPSGKA GAANGFLMEV CVDSVESAVN AERGGADRIE LCSGLSEGGT TPSMGVLQVV KQSVQIPVFV MIRPRGGDFL YSDREIEVMK ADI Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 94%.
Members of the CUT family of copper transporters are associated with copper homeostasis and are involved in the uptake, storage, delivery, and efflux of copper (Gupta et al., 1995 ; Li et al., 2005 ).
Specifications
CUTC | |
Polyclonal | |
Unconjugated | |
CUTC | |
2310039I18Rik; AI326282; CGI-32; Copper homeostasis protein cutC homolog; Cutc; cutC copper transporter; cutC copper transporter homolog; cutC copper transporter homolog (E.coli); RP11-483F11.3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51076 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NTM9 | |
CUTC | |
Recombinant protein corresponding to Human CUTC | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CUTC Polyclonal Antibody, Invitrogen™ > 100 μL; Unconjugated
Research Use Only
Spot an opportunity for improvement?Share a Content Correction