Learn More
Multiroir™ Crystal Polystyrene Transmission Lab Box
Use these boxes for a variety of storage needs. Multiroir™ Crystal Polystyrene Transmission Lab Boxes are ideal for conservation, storage and presentation of products. Boxes are made of crystal polystyrene, a resistant material obtained by polymerization of styrene monomer CH2 = CH-Ph.
Brand:  Multiroir™ 45101
 View more versions of this product
Product Code. 11712944
  • 59.85€ / Pack of 10
Edge
Explore more special offers
Description
Description
  • Transparent, stackable and highly resistant to UV
  • Integrated Label holder
  • Chemically inert plastic
  • Food contact quality
  • Funds and above have male-female reliefs that lock stacking and provide perfect storage stability
  • The tight tolerances of the fitting of the cover on the base ensure a good seal
Specifications
Specifications
0.09 L
Polystyrene
Conservation, Storage and Presentation of Products
10/Pk.
60 mm
45 mm
43 mm
Transparent
Rectangular
Transmission Lab Box
60 L x 50 H mm
55 mm
50 mm
0.03 kg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Multiroir™ Crystal Polystyrene Transmission Lab Box > Capacity: 0.09L; Dimensions: 60L x 50mmH