Learn More
Epredia™ Cryotome™ Cryostat Cryocassettes, 25mm dia. Crosshatched
Enhance operation of Cryotome™ FSE/FE/SME/E Cryostats with Epredia™ Cryotome™ Cryostat Accessories.
Brand: Epredia™ A78930200
This item is not returnable.
View return policy
Description
- Multipositional heat extractor is incorporated on a vertical shaft for even freezing and easy removal from specimen; can be operated from any position on the freeze bar
- Freezing time is shortened if freezing stage and tissue are placed on active Peltier
- Freezing disc is used in conjunction with heat extractor, assisting in the flattening and freezing of tissue samples
Specifications
Glass anti-roll plate | |
Glass Anti-Roll Plate | |
FSE/FE/SME/E |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Epredia™ Cryotome™ Cryostat Cryocassettes, 25mm dia. Crosshatched > Glass anti-roll plate