Learn More
Gonotec™ Osmomat 3000 Single Sample Freezing Point Osmometer
The Gonotec™ Osmomat 3000 Single Sample Freezing Point Osmometer is especially designed for routine measurements in the medical field and is also very suitable for measurements in research and industry.
Brand:  Gonotec™ 32
Product Code. 15104726
  • 13780.00€ / Each
Estimated Shipment: 07-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Determines the total osmolality of aqueous solutions
  • Requires very small sample volumes and can thus be applied for extreme measuring tasks
  • Rapidity allows serial measurements in a very short time
  • Can be controlled easily and comfortably via a touch-screen display
  • Step-by-step user guidance through all measurement functions
  • QM assistance for the laboratory supervisor
  • 2- or 3-point calibration
  • Results are sent to the optional built-in printer in document-ready format
  • For data transfer to a PC it can be connected via USB or RS232
  • Last result remains available for reading even after automatic switching to stand-by mode
  • Robust design of the measurement equipment makes this unit easy to handle and maintain
  • Choice of language menu
  • Automatic calibration by using Gonotec calibration standards
Specifications
Specifications
LCD Touchscreen
General medicine, routine and research, forensic medicine, electron microscopy, physiology, clinical laboratories, paediatrics, urology and more
0 to 3000 mOsmol/kg H2O
1 mOsmol/kg H2O
10°C to 35°C
100/240 V 50/60 Hz 45 VA
By means of two separate peltier cooling systems with heat dissipation by air; crystallization process by means of the tip of a stainless steel needle covered with ice crystals which is automatically controlled
60 sec.
<±2 digit (0...400) mOsmol/kg H2O
< ±0.5% (400...1500) mOsmol/kg H2O
< ±1.0% (1500...3000) mOsmol/kg H2O
50 μL (optional 15 μL)
6.4 kg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Gonotec™ Osmomat 3000 Single Sample Freezing Point Osmometer >