Learn More
Fisherbrand™ Cryogenic Labels for Cryogenic Storage
GREENER_CHOICE
Intended uses of cryostorage with liquid nitrogen, refrigeration and ultra-low temperature freezer applications. Comes in an easy-to-use dispenser box with 20 sheets per pack.
Brand:  Fisherbrand™ 15920E
18.05 EUR valid until 2025-01-31
Use promo code "24160" to get your promotional price.
 View more versions of this product
Product Code. 17807291
  • 23.00€ / Each
Estimated Shipment: 25-03-2025
to see stock.
Explore more special offers
Description
Intended uses of cryostorage with liquid nitrogen, refrigeration and ultra-low temperature freezer applications. Comes in an easy to use dispenser box with 20 sheets per pack.
Ideal for all cryostorage, liquid nitrogen, refrigeration, and ultra-low temperature freezer applications. Suitable for use on microcentrifuge tubes, cryovials, racks, boxes and microtiter plates. Adhesive withstands temperatures from -196° to +121°C (-321° to +250°F)
  • No need to self-laminate
  • Thinner labels allow tubes to fit most storage systems
  • Available in a full range of colors and sizes
  • Guaranteed not to fall off under normal use (provided user instructions are followed)
  • Also offered in laser printer-compatible sheets (see Cat. Nos. 15-930 and 15-940 Series)
Cryostorage, liquid nitrogen, refrigeration, and ultra-low temperature freezer applications
Specifications
Specifications
White
Cryogenic Label
15 and 50 mL tube
0.75 in. dia.
-196°C to +121°C
1.9 cm
Label
Cryostorage, Liquid Nitrogen, refrigeration and ultra-low temperature freezer applications
1000/Roll
-321 to 250°F
0.75 in.
Documents
One moment while we fetch your results.
Product Title
Fisherbrand™ Cryogenic Labels for Cryogenic Storage > 0.75 in. dia.; Roll of 1000; White; Dots