missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Notrax Crossrib Sof-Tred™ Dyna-Shield™ Layer with Side-to-Side Wide Ribs, Black/Yellow
Notrax Crossrib Sof-Tred™ - Exceptional worker comfort due to 12.7 mm thick specially formulated compound. Protective layer of exclusive Dyna-Shield™ extends service life by 50% compared to regular microcellular vinyl.
58.00€ - 154.00€
Specifications
Color | Black/Yellow |
---|---|
Material | Microcellular Vinyl |
Type | Industrial and Commercial Matting |
Product Code | Brand | Length (Metric) | Width (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Width (Metric) | Price | Quantity & Availability | ||||
17792048
|
Notrax
406S0023BY |
91 cm | 60 cm |
58.00€
Each |
Estimated Shipment: 10-01-2025
Log in to see stock. |
||||
17712058
|
Notrax
406S0035BY |
150 cm | 91 cm |
154.00€
Each |
Estimated Shipment: 10-01-2025
Log in to see stock. |
||||
17732048
|
Notrax
406C0024BY |
1830 cm | 60 cm | N/A | |||||
17752048
|
Notrax
406C0036BY |
1830 cm | 91 cm | N/A | |||||
17772048
|
Notrax
406C0048BY |
1830 cm | 122 cm | N/A | |||||
Description
- Exceptional worker comfort due to 127 mm thick specially formulated compound
- Protective layer of exclusive Dyna-Shield™ extends service life by 50% compared to regular microcellular vinyl supported by independent laboratory testing
- Side-to-side wide ribs provide traction and easy to sweep clean
- Available with yellow borders along sides to comply with OSHA code 1910-144
- All 4 sides are bevelled and sealed to minimize tripping hazards
Specifications
Black/Yellow | |
Microcellular Vinyl | |
Industrial and Commercial Matting |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title