missing translation for 'onlineSavingsMsg'
Learn More

Notrax Crossrib Sof-Tred™ Dyna-Shield™ Layer with Side-to-Side Wide Ribs, Black/Yellow

Notrax Crossrib Sof-Tred™ - Exceptional worker comfort due to 12.7 mm thick specially formulated compound. Protective layer of exclusive Dyna-Shield™ extends service life by 50% compared to regular microcellular vinyl.

Brand:  Notrax 406C0048BY

 View more versions of this product

Product Code. 17772048

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Exceptional worker comfort due to 127 mm thick specially formulated compound
  • Protective layer of exclusive Dyna-Shield™ extends service life by 50% compared to regular microcellular vinyl supported by independent laboratory testing
  • Side-to-side wide ribs provide traction and easy to sweep clean
  • Available with yellow borders along sides to comply with OSHA code 1910-144
  • All 4 sides are bevelled and sealed to minimize tripping hazards
Specifications

Specifications

Black/Yellow
1830 cm
Industrial and Commercial Matting
60 ft.
Microcellular Vinyl
122 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.