Learn More
Thermo Scientific™ Crossflow Filtration Accessory Pack
The Crossflow Filtration Accessory Pack features all necessary components including the feed line, return line, filtrate line, and pressure gauge.
Brand: Thermo Scientific™ A57937
31.14 EUR valid until 2025-03-29
Use promo code "24111" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
The Crossflow Filtration Accessory Pack features all necessary components including the feed line, return line, filtrate line, and pressure gauge.
Accessory pack includes:
• Feed line
• Return line
• Filtrate line
• Pressure gauge
Specifications
Not Autoclavable | |
Filtration Systems | |
Non-sterile | |
Each |
Reusable | |
PVC, Polypropylene, Stainless Steel | |
Feed line, return line, filtrate line, and pressure gauge | |
Certified Quality |