missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Costar™ 96-Well, Cell Culture-Treated, U-Shaped-Bottom Microplate
Shop All Corning Products

Click to view available options
Packaging:
Bag of 20; Cs. of 100
Individually wrapped, 50/Cs.
Description
Corning polypropylene microplates offer both small volume and large volume (blocks) well formats to meet assay and storage requirements.
- Flat, round, or V-shaped well bottom
- Features uniform skirt heights for greater robotic gripping surface
- Solvent resistant polypropylene provides compatibility with many common organic solvents (e.g., DMSO, ethanol, methanol)
- RNase-/DNase-free
- Available sterile or nonsterile
Greener Choice Claims
- Environmental benefits include:
- Certifications and Seals of Approval
- Renewable Energy
- Energy Star Certification

Specifications
Specifications
Format | Standard |
Lid | With Lid |
Array | 8 x 12 |
Packaging | Bag of 20; Cs. of 100 |
Color | Clear |
Material | Polystyrene |
No. of Wells | 96 |
Volume (Metric) Working | 75 to 200 μL |
Sterility | Sterile |
Surface Treatment | Cell Culture |
Show More |
Documents
Product Information
Product Information
COR001 Guide selection Culture Cellulaire Corning
File size: 5.5 MB
COR001 Lifescience Guid
File size: 7 MB
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Corning™ Costar™ 96-Well, Cell Culture-Treated, U-Shaped-Bottom Microplate >
Spot an opportunity for improvement?Share a Content Correction