Learn More
Corning™ Nonsterile Polyester Microplate Sealing Tape
Provides well-to-well sealing across entire microplate to eliminate edge effect in sensitive ELISA assays. Corning™ Nonsterile Polyester Microplate Sealing Tape is suitable for sealing tissue culture mmicroplates, for short term storage incubation, for transport and the containment of solutions.
Brand: Corning™ 4612
379.81 EUR valid until 2025-03-31
Use promo code "24073" to get your promotional price.
Description
- Easy application and removal for short- and long-term storage
- Provides tight seal to minimize evaporation and condensation
- Aluminum sealing tape (Cat. Nos. 6569 and 6570) is suitable for use between -80°C and 150°C, is not transparent, and is pierceable.
- Breathable sealing tape (Cat. No. 3345) allows gas exchange across the surface.
- Universal Optical sealing tape (Cat. No. 6575) is suitable for use between -70°C and 100°C, and is transparent.
Specifications
Transparent | |
Non-sterile | |
100/Pk. | |
-40°C to +120°C |
Polyester | |
500/Cs. | |
Microplate Sealing Tape |
Promotions
-
Corning Matrigel
,
Corning Biocoat
,
Corning CoolProduct
,
Corning Media Tech
,
Corning
New Offer Save Now
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
Days
30Hours
19Minutes
21Seconds
10New Offer Save Up to 92%