Learn More
Corning™ Magnetic Stir Bar Retriever
For Use With: Stirring Models
Brand: Corning™ 6970SR
11.48 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
The Corning stir bar retriever contains a strong magnet to remove a stir bar from all glass and plastic containers used in the laboratory. The container must be taken off the magnetic stirrer before using the retriever as the high powered magnet in the stirrer cancels out the one in the retriever. The handle is designed for ergonomic and easy retrieval. -The retriever is available one per case.
Specifications
Stirring Models | |
Accessory, Stir Bar Retriever |
Promotions
-
Corning Matrigel
,
Corning Biocoat
,
Corning CoolProduct
,
Corning Media Tech
,
Corning
-
Multiple Manufacturers
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsPromo Code: 25721Expires: 2025-05-01
New Offer Save Now -
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Days
27Hours
1Minutes
47Seconds
5