missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ 96-Well, Non-Treated, Flat-Bottom, Half-Area Microplate

Color:White, Clear Bottom; Sterility:Sterile; Lid:Without Lid
Brand: Corning™ 3883
This item is not returnable.
View return policy
Description

Specifications
Half-Area | |
8 x 12 | |
Standard automated handling and detection systems | |
96 | |
Sterile | |
Flat Bottom (F-well) | |
100/Cs. | |
100/Cs. |
Without Lid | |
White, Clear Bottom | |
Polystyrene | |
25 to 125 μL | |
Non-treated | |
Suitable for immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface | |
25/Pk. | |
Clear |
COR001 Guide selection Culture Cellulaire Corning
File size: 5.5 MB
COR001 Lifescience Guid
File size: 7 MB
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Corning™ 96-Well, Non-Treated, Flat-Bottom, Half-Area Microplate > White and Clear; Half Area; No; None; Flat
Spot an opportunity for improvement?Share a Content Correction