Learn More
Corning™ 12-Strip, 96-Well, High Binding, Flat-Bottom Microplate
Color:Clear; Sterility:Non-sterile; Lid:Without Lid
Brand: Corning™ 42592
This item is not returnable.
View return policy
Description
Sold 100 per case.
Specifications
Standard | |
8 x 12 | |
Clear | |
Non-sterile | |
Flat Bottom (F-well) |
Without Lid | |
High Binding, 4CAV | |
96 | |
High Binding | |
100/Cs. |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Corning™ 12-Strip, 96-Well, High Binding, Flat-Bottom Microplate > 1 x 8; High binding; Clear; Flat bottom; Without lid; Nonsterile; 4 CAV