Learn More
BINDER™ Copper Interior Fittings
Copper Interior Fittings is a set consisting of 2 shelves and a water pan.
Brand: BINDER™ 8012-1997
Description
Set Includes:
- 2 shelves (all-copper)
- A water pan (copper-plated)
Specifications
Copper interior fittings - Set consisting of 2 shelves (all-copper) and a water pan (copper-plated) | |
Copper | |
2 shelves (all-copper) and a water pan (copper-plated) |
BINDER Chambers CB-S170 | |
Fitting |