missing translation for 'onlineSavingsMsg'
Learn More
Copper(II) chloride, anhydrous, 98% min, Thermo Scientific Chemicals Product Code.: 11381809

Copper(II) chloride, anhydrous, 98% min, Thermo Scientific Chemicals

Product Code. 11381809
1 kg
Click to view available options
Quantity:
50 g
250 g
1 kg
This item is not returnable. View return policy

Product Code. 11381809

Brand: Thermo Scientific Alfa Aesar 012457.A1

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For Isomerization and Cracking Catalyst, Mordant in Dyeing and Printing Fabrics, in Electroplating Baths For Plating Cu On Al

Glassy carbon is widely practiced as an electrode material in electrochemistry, as considerably as for high temperature crucibles and as a part of some prosthetic devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7447-39-4
Molecular Formula CuCl2
MDL Number MFCD00010972

Specifications

Boiling Point 993°C (decomposition)
Melting Point 620°C
Color Brown-Yellow
Physical Form Powder
Quantity 1 kg
Assay Percent Range 98% min.
Solubility Information Soluble in water,alcohol,acetone and hot sulfuric acid.
Formula Weight 134.45
Odor Odorless
Sensitivity Air sensitive; Hygroscopic
Appearance Yellow-brown powder
Density 3.386
Chemical Name or Material Copper(II) chloride, Anhydrous
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW19K056 Certificate TypeCertificate of Analysis Date27/09/2024 Catalog Number012457.30,012457.18,012457.A1
Lot NumberS25K030 Certificate TypeCertificate of Analysis Date03/07/2024 Catalog Number012457.30,012457.18,012457.A1
Lot NumberQ17K034 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number012457.30,012457.18,012457.A1
Lot NumberY03J026 Certificate TypeCertificate of Analysis Date15/11/2023 Catalog Number012457.30,012457.18,012457.A1
Lot Number10237392 Certificate TypeCertificate of Analysis Date11/08/2023 Catalog Number012457.30,012457.18,012457.A1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Copper(II) chloride, anhydrous, 98% min, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.