missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
Boiling Point | 1290°C |
Melting Point | 605°C |
CAS | 7681-65-4 |
Color | Beige |
Physical Form | Solid |
Quantity | 100 g |
Molecular Formula | CuI |
MDL Number | 10978 |
InChI Key | LSXDOTMGLUJQCM-UHFFFAOYSA-M |
SMILES | [Cu+].[I-] |
Show More |
Safety and Handling


Product Identifier
- Copper(I) iodide
- Warning
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Copper(I) Iodide, Technical, SLR, Fisher Chemical >
Spot an opportunity for improvement?Share a Content Correction