Learn More
Memmert™ Convection Oven Stainless Steel Grids
Use these grids with Memmert ovens. Memmert™ Convection Oven Stainless Steel Grids feature sturdy construction and are available in a variety of sizes to fit your needs.
Brand: Memmert™ E20165
92.30 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
- Reinforced stainless steel grid
- For convection ovens
Specifications
Models 110, 160 incubator | |
Grid | |
490 x 370 mm Glass Door | |
Stainless Steel |
Product Suggestions
Customers who viewed this item also viewed
Promotions
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
-
Multiple Manufacturers
Dairy and Plant-Based Products Testing Promotion
Quality testing products for all your needs.Promo Code: 24134Expires: 2025-03-31