Learn More
Grant Instruments Controlled Rate Freeze Thaw (CRFT) with Peli Case

Controlled Rate Freeze Thaw (CRFT) for cryopreservation. Supplied in rigid black peli case. Interchangeable heads must be ordered seperately.
Brand: Grant Instruments CRFT EU PACK
Description
Controlled rate freezer and thawer that maximizes cell survival without using liquid nitrogen.
Applications
- Transgenic embryology research
- Stem cell research
- Clinical and research samples, e.g. lymphocytes and tissue cell lines in conventional cryovials
- Various mammalian cells including cardiomyocytes, adipose, liver and muscle
- Cord blood derived stem cells
- Adherent cells and stem cells in microplates
- Cell suspensions in numbered/barcoded arrays
- System integrations
- Suitable for applications in veterinarian IVF
Key features and benefits
- Accurate and reproducible control of cooling rates and sample temperatures
- A focus on maximising cell survival rates
- Controlled and customisable freezing and thawing
- Linear and non-linear freezing profiles
- Liquid nitrogen, alcohol and cryogen free - no need for 'topping up'
- Interchangeable heads for flexibility (ordered separately)
- Free of charge software allows you to design, track, export and report temperature profiles
- Easy to use and samples can be nucleated/seeded in-situ
- Low running costs: estimated at 1% of liquid nitrogen-controlled rate freezing and runs on a 24 V supply
- Temperature range of -100°C to 30°C
- Servicing and calibration options available
- Command protocols available

Specifications
Cryogenic Freezer/Thawer | |
Plastic | |
Plastic | |
Benchtop | |
White | |
2 Years | |
CRFT | |
571 x 569 x 540 mm | |
Internal Data Logger USB | |
Via Software | |
UPS (As Optional Extra) | |
3.5 A | |
100/240 V | |
170 W | |
Air-cooled | |
44.31 lb. | |
19.62 lb. | |
Interchangeable Heads |
1 | |
Aluminum | |
CE | |
No | |
Vials, Straws, PCR Plates | |
Controlled Thawing | |
No | |
507 x 510 x 498 mm | |
Password Protected Software | |
Yes | |
USB and Software Supplied | |
EU | |
50/60 Hz | |
Up to 55 | |
Helium | |
20.1 kg | |
8.9 kg |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.