missing translation for 'onlineSavingsMsg'
Learn More

VITLAB™ Continuous Bottle-Top Burette

Ensure precise and continuous titration. VITLAB™ Continuous Bottle-Top Buret features a digital control panel with hand wheels to easily control titration rate and calibration. Adjustable components and a double-piston pump ensure precise results

Brand:  VITLAB™ 1620507

 View more versions of this product

Product Code. 10100122

  • 895.77€ / Each
Estimated Shipment: 28-05-2025
to see stock.
Explore more special offers
Add to basket

Récupel-bijdrage uitgesloten
This item is not returnable. View return policy

Description

Description

Chemical volumetric analysis with minute liquid volumes requires substantial concentration. The VITLAB™ Continuous Bottle-Top Buret enables continuous titration up to a concentration of 1 mol/L, which leads to rapid, convenient and accurate results.

Digital display panel simplifies operation
  • Angled display shows titration volume in large, easily read numbers
  • Easy-to-use, medium-specific calibration using the keypad buttons
  • Precise control of the titration rate by rotating two large, easy-grip hand wheels
  • Also includes battery, pause and mode indicators and function keys/buttons
Adjustable, continuous titration control
  • Patented double-piston pump supplies the titration in a continuous, pulse-free manner
  • Telescopic discharge tube can be adjusted (height and length) for use with short or tall bottles
  • Rotates freely 360° around the bottle for optimal orientation of the label
  • Adjustable for various bottle sizes using the corresponding thread adapter
  • Long operating life using two easily-replaced 1.5 V microbatteries
Quality and safety assured
  • Innovative recirculation valve prevents the loss of reagents and reduces splashes
  • Compact design with low center of gravity reduces risk of overturning, especially with smaller bottles
  • Fulfils all requirements for test equipment monitoring without instrument downtime
  • Margins of error are under the DIN EN ISO 8655-3 standard, even for partial volumes
Specifications

Specifications

0.2 %
0.01 (up to 99.99 mL) mL, 0.1 (100 to 999.9 mL) mL
(2) 1.5 V Microbatteries, GL 45 Connecting Threads and GL 32, GL 38 and S*40 Size PP Thread Adapters, Instruction Manual and Quality Certificate, Telescopic Discharge Tube (140-220 mm), Telescopic Filling Tube (200-350 mm)
Continuous Buret
5 mL
Continuous Titration of a Range of Media for Chemical Volumetric Analysis
0.1 %
DIN 12600, DIN EN ISO 8655-3
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
VITLAB™ Continuous Bottle-Top Burette > Capacity: 5 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.