missing translation for 'onlineSavingsMsg'
Learn More
Conductivity Test Solution Potassium Chloride, SI Analytics™ Product Code.: 12037112

Conductivity Test Solution Potassium Chloride, SI Analytics™

Product Code. 12037112
250 mL, Polyethylene Bottle, 0.01 mol/L
Click to view available options
Quantity:
20 mL
250 mL
500 mL
Packaging:
3 x 6 Ampules
Polyethylene Bottle
Concentration:
0.01 mol/L
0.01/0.1/1 mol/L (1.41/12.9/112mS/cm)
0.1 mol/L (12.9mS/cm)
5mS/cm
This item is not returnable. View return policy

Product Code. 12037112

Brand: SI Analytics 285136907

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

SI Analytics™ Conductivity Test Solution Potassium Chloride allows for the determination of a substances ability to conduct electricity.

  • Mixture consisting of water and potassium chloride
  • Store at room temperature
  • The product does not contain any relevant quantities of materials with critical values that have to be monitored at the workplace
  • Product does not present an explosion hazard
  • Do not allow undiluted product or large quantities of it to reach ground water, water course or sewage system
  • Small amounts may be diluted with plenty of water and washed away
TRUSTED_SUSTAINABILITY

Specifications

Packaging Polyethylene Bottle
Quantity 250 mL
Odor Odorless
Density 1g/cm3
Physical Form Fluid
Concentration 0.01 mol/L
Color Colorless
pH 7
Melting Point 0°C
Boiling Point 100°C
Solubility Miscible with water
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Conductivity Test Solution Potassium Chloride, SI Analytics™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.