missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Conductivity Standard, 1413μS/cm, NIST Certified, Chem-Lab
Click to view available options
Quantity:
500 mL
1000 mL
Description
5°C—896μS/cm
10°C—1020μS/cm
15°C—1147μS/cm
20°C—1278μS/cm
25°C—1413μS/cm
30°C—1552μS/cm
35°C—1694μS/cm
40°C—1839μS/cm
Specifications
Specifications
Packaging | Polyethylene |
Quantity | 1000 mL |
Density | 1.01g/mL |
Conductivity | 1413μS/cm |
Concentration | 0.01 M |
IUPAC Name | potassium chloride |
Formula Weight | 74.56g/mol |
Chemical Name or Material | Potassium Chloride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Conductivity Standard, 1413μS/cm, NIST Certified, Chem-Lab >
Spot an opportunity for improvement?Share a Content Correction