missing translation for 'onlineSavingsMsg'
Learn More
1413 μS/cm Conductivity Standard Solution Sachets, Hanna Instruments™ Product Code.: 11616990

1413 μS/cm Conductivity Standard Solution Sachets, Hanna Instruments™

Product Code. 11616990
500 mL, Light Block Bottles
Click to view available options
Quantity:
500 mL
25 x 20 mL
Packaging:
Light Block Bottles
Sachets
This item is not returnable. View return policy

Product Code. 11616990

Brand: Hanna Instruments

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Premium quality 1,413 μS/cm conductivity solution that is NIST traceable and supplied with a Certificate of Analysis that is ideal for users that do not calibrate frequently.

  • Premium quality 1,413 μS/cm conductivity solution that is NIST traceable and supplied with a Certificate of Analysis
  • Temperature chart of the actual μS/cm value at various temperatures printed on each bottle
  • 1,413 μS/cm conductivity solution standardized with a conductivity meter that has been calibrated with potassium chloride NIST 2202 SRM (Standard Reference Material)  
  • Deionized water for analytical use in accordance with ISO 3696/BS3978
  • Additional equipment used in preparation of the solutions include certified weight-checked balances, Class A glassware and certified thermometers
  • Ideal for users that do not calibrate frequently
  • Ideal for field use due to portability

Specifications

Packaging Light Block Bottles
Quantity 500 mL
Color Colorless
Odor Odorless
pH 7
Solubility Soluble
Physical Form Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1413 μS/cm Conductivity Standard Solution Sachets, Hanna Instruments™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.