Learn More
Lenz Laborglasintrumente Condensers acc. to Allihn, for Extractors, with Olives
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
117.00€ - 125.00€
Product Code | Brand | Standard Taper Joint No | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint No | Price | Quantity & Availability | ||||||
10306171
|
Lenz Laborglasintrumente
05226004 |
NS 29/32 |
117.00€
Each |
Estimated Shipment: 12-05-2025
Log in to see stock. |
||||||
10244101
|
Lenz Laborglasintrumente
05226005 |
NS 34/35 |
125.00€
Each |
Estimated Shipment: 03-06-2025
Log in to see stock. |
||||||
Description
- Allihn condensers for condensing vapours
- With NS cone
- Water connection: olives
- Made of DURAN™ tubing
Specifications
Duran Glass |
Product Title
Lenz Laborglasintrumente Condensers acc. to Allihn, for Extractors, with Olives