missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Concentrator Plus
Eppendorf Concentrator plus was designed to deliver an ergonomic, intuitive and durable solution that fits comfortably right on your lab bench.
Brand: Eppendorf™ 5305000304
8862.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description

Specifications
50/60 Hz | |
20 Mbar | |
Concentrators | |
Concentrator Plus Complete | |
230 mm | |
350 W | |
58 x 33 x 29 cm (12.6 x 20.9 x 11.7 in.) |
With Rotor F-45-48-11 | |
Integrated diaphragm vacuum pump | |
230 V | |
230 V, 50/60 Hz (EU) | |
1400 rpm | |
330 mm | |
Rotor F-45-48-1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Eppendorf™ Concentrator Plus > Model: Concentrator plus; Includes: with Rotor F-45-48-11; Pump type: with integrated diaphragm vacuum pump;Electrical Requirements: 230V/50–60Hz (EU);
Spot an opportunity for improvement?Share a Content Correction