missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
For few parameters such as nitrite, which are subject to a quick conversion, freshly prepared control standards of stock solutions versus the multi-parameter standard checks are mandatory. Stock solutions are also more cost-effective than the already confectioned CombiChecks™. With stock solutions, the correct dilution must be self-calculated and conducted. The use of standards is supported via AQA functions in the photoLab™ photometers.
- Free chlorine
- Bound chlorine
- Formaldehyde
- Hydrazine
- Hydrogen peroxide
- Hydrogen sulfide
- Phenol
- Silicon
- Sulfide
- Sulfite
- Anionic surfactants

Specifications
Specifications
Quantity | 100 mL |
Physical Form | Liquid |
Concentration | 100 mg/L |
Color | Green/White |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
COD Standard Solution, 100mg/L, WTW™ >
Spot an opportunity for improvement?Share a Content Correction