Learn More
behr Labor-Technik™ COD Digestor System
Capable of simultaneous EPA, ISO, or DIN method digestion of up to 8/12 COD samples. behr Labor-Technik™ COD Digestor System is a workstation comprising all necessary components for COD sample digestion.
7420.00€ - 8690.00€
Specifications
Product Type COD Digestor
Includes Time/temperature regulator, heating block, holding frame, cooling bath, sample vessels, air-cooled condensers, condenser stand
View More Specs
Products 2
Product Code Brand No. of Places Price Quantity & Availability  
Product Code Brand No. of Places Price Quantity & Availability  
10074130
View accessory products View Documents
behr Labor-Technik™
B00218405
8
7420.00€
Each
Estimated Shipment: 16-01-2025
to see stock.
Add to basket
10772001
View accessory products View Documents
behr Labor-Technik™
B00218406
12
8690.00€
Each
Estimated Shipment: 11-12-2024
to see stock.
Add to basket
Description
Description
  • Systems consist of the following components:
    • Microprocessor-controlled TRS 200 time/temperature regulator with COD program
    • Precision heating block CSB/E for RG2 vessels
    • Holding frame E/B for RG2 sample vessels
    • Cooling bath KW/N with mounting for E/B frame
    • Sample vessels RG 2
    • Air-cooled condensers LK 1
    • Condenser stand LS for LK condensers
Specifications
Specifications
COD Digestor
Time/temperature regulator, heating block, holding frame, cooling bath, sample vessels, air-cooled condensers, condenser stand