Learn More
behr Labor-Technik™ COD Digestor System
Capable of simultaneous EPA, ISO, or DIN method digestion of up to 8/12 COD samples. behr Labor-Technik™ COD Digestor System is a workstation comprising all necessary components for COD sample digestion.
Brand:  behr Labor-Technik™ B00218405
 View more versions of this product
Product Code. 10074130
  • 7420.00€ / Each
Estimated Shipment: 16-01-2025
to see stock.
Explore more special offers
Add to basket
Récupel-bijdrage uitgesloten
Description
Description
  • Systems consist of the following components:
    • Microprocessor-controlled TRS 200 time/temperature regulator with COD program
    • Precision heating block CSB/E for RG2 vessels
    • Holding frame E/B for RG2 sample vessels
    • Cooling bath KW/N with mounting for E/B frame
    • Sample vessels RG 2
    • Air-cooled condensers LK 1
    • Condenser stand LS for LK condensers
Specifications
Specifications
COD Digestor
Time/temperature regulator, heating block, holding frame, cooling bath, sample vessels, air-cooled condensers, condenser stand
8
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products