Learn More
BINDER™ Coated Heat Exchanger
Heat exchanger that is coated in plastic to prevent damage.
950.00€ - 1750.00€
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | ||||||
30238286
|
BINDER™
8012-2462 |
KB PRO 260 |
950.00€
Each |
Estimated Shipment: 22-04-2025
Log in to see stock. |
||||||
30238248
|
BINDER™
8012-2463 |
KB PRO 470 |
950.00€
Each |
Estimated Shipment: 22-04-2025
Log in to see stock. |
||||||
30238208
|
BINDER™
8012-2464 |
KB PRO 720, KB PRO 1060 |
950.00€
Each |
Estimated Shipment: 22-04-2025
Log in to see stock. |
||||||
30238303
|
BINDER™
8012-2580 |
KBF PRO 130 |
950.00€
Each |
Estimated Shipment: 22-04-2025
Log in to see stock. |
||||||
30238380
|
BINDER™
8012-2581 |
KB PRO 1600 |
1750.00€
Each |
Estimated Shipment: 22-04-2025
Log in to see stock. |
||||||
30238376
|
BINDER™
8012-2583 |
KBF PRO 260 |
950.00€
Each |
Estimated Shipment: 22-04-2025
Log in to see stock. |
||||||
30238318
|
BINDER™
8012-2584 |
KBF PRO 470 |
950.00€
Each |
Estimated Shipment: 22-04-2025
Log in to see stock. |
||||||
30238329
|
BINDER™
8012-2585 |
KBF PRO 720, KBF PRO 1060 |
950.00€
Each |
Estimated Shipment: 22-04-2025
Log in to see stock. |
||||||
30238287
|
BINDER™
8012-2586 |
KBF PRO 1600 |
1750.00€
Each |
Estimated Shipment: 22-04-2025
Log in to see stock. |
||||||
Description
Heat exchanger that is coated in plastic to prevent damage to the heat exchanger caused by corrosive sample material.
Specifications
Aluminum | |
Heat Exchanger |
Product Title
BINDER™ Coated Heat Exchanger