Learn More
Brand™ Class B AR-Glass/Soda Lime Glass SilberEterna™ Volumetric Bulb Pipet
These bulb pipettes provide the highest accuracy, maintained by strict statistical quality controls.
Brand:  Brand™ 29519
 View more versions of this product
Product Code. 11848301
Description
Description
  • Acc. to DIN EN ISO 648
  • 0.5 to 2mL capacity bulbs provided in a pack of 12
  • 3 to 100mL bulbs provided in a pack of 6
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Autoclavable
DIN EN ISO 648
AR-GLAS, Soda Lime Glass
Amber
±0.12 mL
585 mm
Yes
B
Yes
6 Pack
100 mL
Volumetric Bulb Pipette
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Brand™ Class B AR-Glass/Soda Lime Glass SilberEterna™ Volumetric Bulb Pipet > Capacity: 100mL