missing translation for 'onlineSavingsMsg'
Learn More

CIQ, Tocris Bioscience™

Product Code. 30001344
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potentiator of NMDA receptors containing GluN2C/GluN2D

Subunit-selective potentiator of NMDA receptors containing the GluN2C (formally NR2C) or GluN2D (formally NR2D) subunit. Increases channel opening efficiency and enhances NMDA receptor responses.

Please refer to IUPHAR Guide to Pharmacology for the most recent naming conventions.

Chemical Identifiers

CAS 486427-17-2
Molecular Formula C26H26ClNO5
Molecular Weight (g/mol) 467.946
InChI Key VYMILMYEENZHAR-UHFFFAOYSA-N
PubChem CID 4231127
IUPAC Name (3-chlorophenyl)-[6,7-dimethoxy-1-[(4-methoxyphenoxy)methyl]-3,4-dihydro-1H-isoquinolin-2-yl]methanone
SMILES COC1=CC=C(C=C1)OCC2C3=CC(=C(C=C3CCN2C(=O)C4=CC(=CC=C4)Cl)OC)OC

Specifications

Quantity 10 mg
Synonym ciq, 3-chlorophenyl 6,7-dimethoxy-1-4-methoxyphenoxy methyl-3,4-dihydroisoquinolin-2 1h-yl methanone, 3-chlorophenyl 3,4-dihydro-6,7-dimethoxy-1-4-methoxyphenoxy methyl-2 1h-isoquinolinyl methanone, 3-chlorophenyl 6,7-dimethoxy-1-4-methoxyphenoxy methyl-3,4-dihydroisoquinolin-2 1 h-yl methanone, 3-chlorophenyl-1r-6,7-dimethoxy-1-4-methoxyphenoxy methyl-3,4-dihydro-1h-isoquinolin-2-yl me, 3-chlorophenyl-6,7-dimethoxy-1-4-methoxyphenoxy methyl-3,4-dihydro-1h-isoquinolin-2-yl methanone, 3-chlorophenyl 3,4-dihydro-6,7-dimethoxy-1-4-methoxyphenoxy methyl-2 1h-isoquinolinyl-methanone, 2-3-chlorobenzoyl-6,7-dimethoxy-1-4-methoxyphenoxy methyl-1,2,3,4-tetrahydroisoquinoline
Formula Weight 467.94
Percent Purity >98%
Chemical Name or Material CIQ
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'