missing translation for 'onlineSavingsMsg'
Learn More

Proteintech ChromoTek mNeonGreen-Trap Magnetic Agarose

The ChromoTek mNeonGreen-Trap Magnetic Agarose is an affinity resin for IP of mNeonGreen-fusion proteins.

238.00€ - 5899.00€

Specifications

Content And Storage Upon receipt store at 4°C. Do not freeze.
For Use With (Application) IP, CoIP, ChIP, RIP
Target mNeonGreen, Branchiostoma lanceolatum
Label or Dye Magnetic Agarose
Species Alpaca
View More Specs

Products 5
Product Code Brand Sufficient For Price Quantity & Availability  
Product Code Brand Sufficient For Price Quantity & Availability  
17303403
View Documents
Proteintech
ntma-200
200 Reactions N/A
N/A
17313403
View Documents
Proteintech
NTMA-400
400 Reactions N/A
N/A
17373393
View Documents
Proteintech
ntma-10
10 Reactions N/A
N/A
17383393
View Documents
Proteintech
ntma-100
100 Reactions N/A
N/A
17393393
View Documents
Proteintech
ntma-20
20 Reactions N/A
N/A
Description

Description

  • Consists of a mNeonGreen Nanobody/ VHH coupled to magnetic agarose beads
Specifications

Specifications

Upon receipt store at 4°C. Do not freeze.
mNeonGreen, Branchiostoma lanceolatum
Alpaca
IP, CoIP, ChIP, RIP
Magnetic Agarose
Videos
SDS
Documents

Documents

Product Certifications