Learn More
Proteintech ChromoTek mNeonGreen-Trap Magnetic Agarose
The ChromoTek mNeonGreen-Trap Magnetic Agarose is an affinity resin for IP of mNeonGreen-fusion proteins.
Brand:  Proteintech ntma-10
 View more versions of this product
Product Code. 17373393
  • 238.00€ / 10 reactions
In Stock
to see stock.
Explore more special offers
Description
  • Consists of a mNeonGreen Nanobody/ VHH coupled to magnetic agarose beads
Specifications
Specifications
Upon receipt store at 4°C. Do not freeze.
mNeonGreen, Branchiostoma lanceolatum
Alpaca
IP, CoIP, ChIP, RIP
Magnetic Agarose
10 Reactions