missing translation for 'onlineSavingsMsg'
Learn More
Chromosulfuric acid, Thermo Scientific Chemicals Product Code.: 10511574

Chromosulfuric acid, Thermo Scientific Chemicals

Product Code. 10511574
2.5 L, Glass bottle
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 10511574

Brand: Thermo Scientific Acros 295510025

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65272-71-1
Molecular Formula Cr2K2O7·H2O4S
Molecular Weight (g/mol) 200.237
MDL Number MFCD00010952
InChI Key IFMFTIBWWVBKOC-UHFFFAOYSA-L
Synonym absorbency linearity solution #2 24.28 mg/l inverted exclamation marka in hclo4, 25 ml
PubChem CID 131664149
IUPAC Name dipotassium;chromium;dihydroxide;dihydrate
SMILES O.O.[OH-].[OH-].[K+].[K+].[Cr]

Specifications

Color Brown to Brown-Red
Boiling Point 330.0°C
Density 1.8400g/mL
Packaging Glass bottle
Specific Gravity 1.84
Quantity 2.5 L
Physical Form Liquid
Solubility Information Solubility in water: soluble in water (caution ! development of heat)
Chemical Name or Material Chromosulfuric acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0418854 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number295510025,295510010
Lot NumberA0393063 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number295510025,295510010
Lot NumberA0403865 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number295510025,295510010
Lot NumberA0410437 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number295510025,295510010
Lot NumberA0413709 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number295510025,295510010

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Chromosulfuric acid, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.