Learn More
ChromaCare™ LC-MS Biologics Flush Solution, Thermo Scientific™
Flush solution for cleaning blood, plasma, and urine samples from the chromatograph.
Brand: Fisher Chemical MB124-1
Description
- Suitable for flushing difficult to clean samples such as blood, plasma, and urine after vitamin D, hormone or drugs of abuse testing
- Pre-mixed for ease-of-use
- Packaged in borosilicate glass bottles to prevent metal adduct formation
- Bottles can be used directly on instrument

Specifications
2-Propanol (45%), Acetonitrile (45%), and Acetone (10%) Solution | |
Mobile-phase solvent blend | |
Colorless | |
0.787 g/cm3 | |
1 L | |
Liquid | |
Pass Test (By GC-FID) | |
45% 2-Propanol: 45% Acetonitrile: 10% Acetone |
6 to 7 | |
≤1.0 AU at 210nm, ≤1.0 AU at 220nm | |
67-64-1, 75-05-8 | |
75°C | |
Borosilicate Glass Bottle | |
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
LC-MS |
Product Suggestions
Customers who viewed this item also viewed.
Safety and Handling
- 45% Isopropyl Alcohol + 45% Acetonitrile + 10% Acetone
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
- MIXTURE LIST-Contains : Acetonitrile, Isopropyl alcohol, Acetone
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
198727 | Certificate of Analysis | 15/11/2023 | MB124-1,MB124-212 |
198726 | Certificate of Analysis | 15/11/2023 | MB124-1,MB124-212 |
201326 | Certificate of Analysis | 13/11/2023 | MB124-1,MB124-212 |
201327 | Certificate of Analysis | 13/11/2023 | MB124-1,MB124-212 |
203711 | Certificate of Analysis | 13/11/2023 | MB124-1,MB124-212 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only