missing translation for 'onlineSavingsMsg'
Learn More
Chloride, Ion chromatography standard solution, Specpure™ Cl∣- 1000μg/mL, Thermo Scientific Chemicals Product Code.: 11405020

Chloride, Ion chromatography standard solution, Specpure™ Cl∣- 1000μg/mL, Thermo Scientific Chemicals

Product Code. 11405020
500 mL
Click to view available options
Quantity:
100 mL
500 mL
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price

Product Code. 11405020

Brand: Thermo Scientific Alfa Aesar 035551.AP

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price

Chloride, Ion chromatography standard solution is used as a standard solution in analytical chemistry and in ion chromatography. It is used for calibration of ion chromatography and other analytical techniques.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00143949

Specifications

Quantity 500 mL
Physical Form Liquid
Solubility Immiscible with water
Grade Specpure™
Chemical Name or Material Chloride Ion chromatography standard solution, Cl- 1000μg/mL

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1542174 Certificate TypeCertificate of Analysis Date17/03/2025 Catalog Number035551.AP,035551.AE
Lot Number1527945 Certificate TypeCertificate of Analysis Date11/02/2025 Catalog Number035551.AP,035551.AE
Lot Number1521539 Certificate TypeCertificate of Analysis Date15/01/2025 Catalog Number035551.AP,035551.AE
Lot Number1521756 Certificate TypeCertificate of Analysis Date15/01/2025 Catalog Number035551.AP,035551.AE
Lot Number1514399 Certificate TypeCertificate of Analysis Date20/12/2024 Catalog Number035551.AP,035551.AE

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Chloride, Ion chromatography standard solution, Specpure™ Cl∣- 1000μg/mL, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.