Learn More
CHEMetrics Cyanide (Free) CHEMets Refill
Refill ampules for CHEMets cyanide test kit K-3810 for the analysis of cyanide (free)
Brand: CHEMetrics R-3810
Description
- 30 ampules
- Method: Isonicotonic Barbituric Acid
- Range, 0 to 0.1 and 0.1 to 1 ppm
- MDL 0.005 ppm
Specifications
Cyanide, Free | |
30 self-filling CHEMets ampoules to refill kit K-3810 | |
CHEMets Cyanide Refill |
C-3801 and C-3810 comparators | |
0 to 0.1 ppm and 0.1 to 1 ppm |
Safety and Handling
- CHEMets cyanide refills
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
- MIXTURE LIST-Contains : barbituric acid, sodium hydroxide, isonicotinic acid, potassium phosphate, monobasic
Your input is important to us. Please complete this form to provide feedback related to the content on this product.