Learn More
Masterflex™ Chem-Durance Bio L/S™ Two-Stop Pump Tubing
Excellent chemical resistance and long pumping life
Brand: Masterflex™ 96117-13
This item is not returnable.
View return policy
Description
- Meets USP Class VI requirements
- Flexible outer jacket acts as an insulating layer to absorb and dissipate heat from the rollers - extending the life of the tubing
- Plasticizer-free, smooth bore liner is hydrophobic and resists the absorption/adsorption of aqueous fluids
Specifications
0.8 mm | |
-60°C | |
13 | |
1/16 in. | |
40 psi | |
74°C |
Chem-Durance Bio | |
1 Each | |
0.06 to 36 mL/min. | |
8.8 m | |
660 mm | |
2-stop Tubing |
Product Content Correction
Product Title
Masterflex™ Chem-Durance Bio L/S™ Two-Stop Pump Tubing > Tubing Size: 13