Learn More
Hanna Instruments™ Checkfridge™ Remote Sensor Thermometer
Designed to monitor temperatures of fridges, freezers and other appliances. Hanna Instruments™ Checkfridge™ Remote Sensor Thermometer is a magnetic, continuous readout digital thermometer fitted with a stainless steel-tipped probe on a 1m cable. Unique CAL Check™ feature provides internal calibration of the device with the flick of a switch, alleviating the time preparing an ice bath.
Brand: Hanna Instruments™ HI147-00
Description
A magnetic, continuous readout digital thermometer fitted with a stainless steel-tipped probe on a 1m cable. Unique CAL Check™ feature allows for an internal calibration of the device with the flick of a switch, thereby alleviating the time preparing an ice bath.- Designed to monitor temperatures of fridges, freezers and other appliances.
- Two strong magnets are incorporated on the back of the meter that enable it to be securely fixed to any steel surface.
- The sensor can be used for either air or product temperature monitoring.
- A built-in calibration check device is activated at the touch of a button to ensure accurate readings at all times.
- 1 x 1.5V AAA Battery included.
Specifications
2 Points | |
AAA battery | |
Stainless Steel General Purpose | |
150°C | |
50°C to 150°C | |
±0.3°C (−20 to 90°C) ±0.5°C (outside) |
5 mm | |
1 m | |
0.1° C | |
50°C | |
Yes |