Learn More
Eppendorf™ Charger Shell 2 for one Multipipette™ E3/E3x or Multipipette™ stream/Xstream
Holder and charger designed for installation in Pipette Carousel 2
Brand: Eppendorf™ 3116603003
57.57 EUR valid until 2025-03-30
Use promo code "24134" to get your promotional price.
Description
- Carries and charges one Multipette™E3/E#x or Multipette™stream/Xstream
- Designed for installation in Pipette Carousel 2
Specifications
Holding/charging pipette | |
For one Multipipette™ E3/E3x or Multipipette™ stream/Xstream, Carousel 2 |
Promotions
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
New Offer Save Up to 92% -
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
-
Multiple Manufacturers
Dairy and Plant-Based Products Testing Promotion
Quality testing products for all your needs.Promo Code: 24134Expires: 2025-03-31