missing translation for 'onlineSavingsMsg'
Learn More
Cetyltrimethylammonium bromide, ∽98%, Molecular biology reagent grade, MP Biomedicals™ Product Code.: 11486461
compliance-icons compliance-icons compliance-icons compliance-icons

Cetyltrimethylammonium bromide, ∽98%, Molecular biology reagent grade, MP Biomedicals™

Product Code. 11486461 Shop All MP Biomedicals Products
500 g
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
This item is not returnable. View return policy

Product Code. 11486461

Brand: MP Biomedicals 0219400490

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 57-09-0
Molecular Formula C19H42BrN
Molecular Weight (g/mol) 364.46
MDL Number MFCD00011772
InChI Key LZZYPRNAOMGNLH-UHFFFAOYSA-M
Synonym cetrimonium bromide, cetyltrimethylammonium bromide, hexadecyltrimethylammonium bromide, ctab, centimide, cetab, bromat, ctmab, cee dee, cetavlon
PubChem CID 5974
ChEBI CHEBI:3567
IUPAC Name hexadecyl(trimethyl)azanium;bromide
SMILES [Br-].CCCCCCCCCCCCCCCC[N+](C)(C)C

Specifications

Color White
Quantity 500 g
Formula Weight 364.5
Percent Purity ∼98%
Grade Molecular Biology Reagent
Physical Form Powder
Chemical Name or Material Cetyltrimethylammonium bromide
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cetyltrimethylammonium Bromide, molecular biology reagent
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : cetrimonium bromide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cetyltrimethylammonium bromide, ∽98%, Molecular biology reagent grade, MP Biomedicals™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.