Learn More
Thermo Scientific™ Ceramic Homogenizers for QuEChERS Kits
Use Thermo Scientific™ Ceramic Homogenizers to grind and homogenize samples for your QuEChERS sample preparation workflow.
Brand: Thermo Scientific™ 60106-CH-15
46.59 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
Use Thermo Scientific™ Ceramic Homogenizers to help accelerate the homogenization process and increase recovery and reproducibility during your QuEChERS (Quick, Easy, Cheap, Effective, Rugged and Safe) sample preparation workflow. These homognizers can be used with all of our QuEChERS Extraction and Clean-Up kits to supplement Original 2003 Method, AOAC 2007.01 Method, EN 15662 Method, and NY/T 1380-2007 Method procedures.
Specifications
Ceramic Homogenizers for 15 mL Tubes, 100/Pk. | |
15 mL Tubes | |
Ceramic Homogenizer |
Promotions
-
Multiple Manufacturers
Dairy and Plant-Based Products Testing Promotion
Quality testing products for all your needs.Promo Code: 24134Expires: 2025-03-31
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30