Learn More
Labconco™ CentriVap™ Complete Concentrator
BIOPHARMA
Comes with everything needed to start processing
Brand:  Labconco™ 7315030
 View more versions of this product
Product Code. 15162995
  • 22560.00€ / Each
Estimated Shipment: 15-09-2025
to see stock.
Explore more special offers
Add to basket
Récupel-bijdrage uitgesloten
Description
Description
Comes with everything needed to start processing
Uses a combination of centrifugal force, vacuum and heat to speed evaporation.
Includes:
  • Vacuum Concentrator
  • -50°C Cold Trap
  • Aluminum rotor holds 40 microcentrifuge tubes and 12-13mm tubes
  • Chemical-resistant diaphragm vacuum pump
  • CentriZap Strobe Light allows user to view samples while rotor is spinning and lid is closed
Molecular biology, proteomics, genomics, cell biology, microbiology, biochemistry, drug discovery and analytical chemistry
Specifications
Specifications
Centrivap
International
Heat Boost Not Included
230 V 50 Hz 6 A
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Labconco™ CentriVap™ Complete Concentrator > International; Heat boost not included