missing translation for 'onlineSavingsMsg'
Learn More

CD44 Antibody (Hermes-1) - Azide and BSA Free, Novus Biologicals™

Rat Monoclonal Antibody has been used in 4 publications

200.00€ - 506.00€

Specifications

Antigen CD44
Clone Hermes-1
Concentration 1 mg/mL
Dilution Western Blot 1:1 - 1:100, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:200, Immunoprecipitation 1:10 - 1:50, CyTOF-ready, Neutralization
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, CyTOF, Neutralization
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18486301
View Documents
Novus Biologicals
NBP2-22530-50ug
50 ug
200.00€
50µg
Please to purchase this item. Need a web account? Register with us today!
18716813
View Documents
Novus Biologicals
NBP2-22530
506.00€
200µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD44 Monoclonal specifically detects CD44 in Human, Mouse, Bovine, Goat, Rabbit, Sheep samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, CyTOF-ready, Block/Neutralize.
Specifications
Show More
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
CD44 Antibody (Hermes-1) - Azide and BSA Free, Novus Biologicals™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.