Learn More
WTW™ Case Set for WTW™ MultiLine™ Multiparameter IDS System
Brand:  WTW™ 2F0004
296.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 15355251
  • 312.00€ / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Specifications
Armoring SM Pro, 3 IDS sensors (large field case), buffer STP 4 and STP 7, stand, beaker, conductivity standard 1413μS/cm at 25°C
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
WTW™ Case Set for WTW™ MultiLine™ Multiparameter IDS System > Includes: Armoring SM Pro, 3 IDS sensors (large field case), buffer STP 4 and STP 7, stand, beaker, conductivity standard 1413μS/cm at 25C