missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbonic Anhydrase IX/CA9 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
292.00€ - 570.00€
Specifications
Antigen | Carbonic Anhydrase IX/CA9 |
---|---|
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1 - 3 ug/ml, Simple Western 1:50, Flow Cytometry 1:1000, ELISA reported in scientific literature (PMID 19963243), Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 2 - 5 ug/ml, Immunoprecipitation 1:10 - 1:500. Use reported in multiple pieces of scientific literature, Immunohistochemistry-Paraffin 1:200 - 1:500, Immunohistochemistry-Frozen 1:200 - 1:500, Immunoblotting, Gel Super Shift Assays, Proximity Ligation Assay, Microarray reported in scientific literature (PMID 31955345), Chromatin Immunoprecipitation (ChIP) 1:10-1:500, Dual RNAscope ISH-IHC 1:1000 |
Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunoblot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), In Situ Hybridization (ISH), In situ PLA |
Classification | Polyclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18785623
|
Novus Biologicals
NB100-417SS |
0.025 mL |
292.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18797463
|
Novus Biologicals
NB100-417 |
570.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
Carbonic Anhydrase IX/CA9 Polyclonal specifically detects Carbonic Anhydrase IX/CA9 in Human, Mouse, Rat, Plant samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Gel Super Shift Assays, Proximity Ligation Assay, Microarray, Dual RNAscope ISH-IHC.Specifications
Carbonic Anhydrase IX/CA9 | |
Western Blot 1 - 3 ug/ml, Simple Western 1:50, Flow Cytometry 1:1000, ELISA reported in scientific literature (PMID 19963243), Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 2 - 5 ug/ml, Immunoprecipitation 1:10 - 1:500. Use reported in multiple pieces of scientific literature, Immunohistochemistry-Paraffin 1:200 - 1:500, Immunohistochemistry-Frozen 1:200 - 1:500, Immunoblotting, Gel Super Shift Assays, Proximity Ligation Assay, Microarray reported in scientific literature (PMID 31955345), Chromatin Immunoprecipitation (ChIP) 1:10-1:500, Dual RNAscope ISH-IHC 1:1000 | |
Polyclonal | |
Rabbit | |
Cancer, Cellular Markers, HIF Target Genes, Hypoxia, Lipid and Metabolism, Signal Transduction | |
PBS with 0.02% Sodium Azide | |
CA-IX, CAIXcarbonic anhydrase 9, Carbonate dehydratase IX, carbonic anhydrase IXpMW1, carbonic dehydratase, EC 4.2.1.1, G250, Membrane antigen MN, MNRCC-associated antigen G250, P54/58N, RCC-associated protein G250, Renal cell carcinoma-associated antigen G250 | |
CA9 | |
IgG | |
Affinity Purified | |
Human, dog, rat and mouse (PMID 22842475). |
1.0 mg/mL | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunoblot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), In Situ Hybridization (ISH), In situ PLA | |
Unconjugated | |
RUO | |
Human, Mouse, Rat, Canine, Plant | |
Q16790 | |
768 | |
A synthetic peptide made to a C-terminal sequence of human Carbonic Anhydrase IX (within residues 400-459) [UniProt Q16790] | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. | |
55 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Carbonic Anhydrase IX/CA9 Antibody - BSA Free, Novus Biologicals™